missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human/Mouse/Rat BMP-2 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand: R&D Systems™ 355-BM-500/CF
3925.60 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
Measured by its ability to induce alkaline phosphatase production by ATDC5 mouse chondrogenic cells. Binnerts, M.E. et al. (2004) Biochem. Biophys. Res. Commun. 315(2):272. The ED50 for this effect is 40-200 ng/mL.Specifications
P12643 | |
Lyophilized from a 0.2 μm filtered solution in Glycine, Sucrose, Tween™ 80 and Glutamic Acid. | |
13 kDa (monomer) | |
500 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. | |
Disulfide-linked homodimer | |
BDA2, BMP2, BMP-2, BMP-2A, BMP2ABone morphogenetic protein 2A, bone morphogenetic protein 2, SSFSC | |
Unconjugated | |
Human, Mouse, Rat | |
Reconstitute at 100 to 200 μg/mL in sterile 4 mM HCI. |
Bioactivity | |
650 | |
BMP-2 | |
Chinese Hamster Ovary cell line,CHO-derived BMP-2 protein Gln283-Arg396 | |
RUO | |
<0.10 EU per 1 μg of the protein by the LAL method. | |
BMP2 | |
BMP-2 | |
Recombinant | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |