missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human LRP-1 Cluster II Fc Chimera Protein
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity
Brand: R&D Systems™ 2368-L2-050
This item is not returnable.
View return policy
Description
The Recombinant Human LRP-1 Cluster II Fc Chimera Protein is derived from NS0. The Recombinant Human LRP-1 Cluster II Fc Chimera Protein has been validated for the following applications: Binding Activity.Specifications
Lyophilized from a 0.2μm filtered solution in PBS. | |
68.1 kDa | |
50 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
<0.1 EU per 1μg of the protein by the LAL method. | |
Recombinant |
4035 | |
LRP-1 Cluster II Fc Chimera | |
Mouse myeloma cell line,NS0-derived human LRP-1 Cluster II protein, (N-terminus) Human LRP-1C2 (Arg786-Leu1165) Accession &Num; Q07954 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) | |
Disulfide-linked homodimer | |
Unconjugated | |
90%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction