missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IYD Protein
A cDNA sequence encoding the IYD was constructed and used to recombinantly synthesize the protein.
137.00€ - 4090.00€
Specifications
Name | IYD Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15930519
|
enQuireBio™
QP12467-5ug |
5 μg |
137.00€
5µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15920519
|
enQuireBio™
QP12467-20ug |
20 μg |
212.00€
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15910519
|
enQuireBio™
QP12467-1mg |
1 mg |
4090.00€
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
IYD Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
IYD protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.4M UREA and 10% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSDRSMEKK KGEPRTRAEA RPWVDEDLKD SSDLHQAEED ADEWQESEEN VEHIPFSHNH YPEKEMVKRS QEFYELLNKR RSVRFISNEQ VPMEVIDNVI RTAGTAPSGA HTEPWTFVVV KDPDVKHKIR KIIEEEEEIN YMKRMGHRWV TDLKKLRTNW IKEYLDTAPI LILIFKQVHG FAANGKKKVH YYNE | |
Greater than 90.0% as determined by SDS-PAGE. |