missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IMPDH1 Protein
A cDNA sequence encoding the IMPDH1 was constructed and used to recombinantly synthesize the protein.
137.00€ - 7875.00€
Specifications
Gene ID (Entrez) | 3614 |
---|---|
Name | IMPDH1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | IMPDH1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15920419
|
enQuireBio™
QP12437-2ug |
2 μg |
137.00€
2µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15900419
|
enQuireBio™
QP12437-10ug |
10 μg |
212.00€
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15910419
|
enQuireBio™
QP12437-1mg |
1 mg |
7875.00€
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
3614 | |
Research Use Only | |
IMPDH1 | |
Human | |
His | |
IMPDH1 Human solution containing 20mM Tris-HCl pH 8.0, 1mM DTT and 20% glycerol. |
IMPDH1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MADYLISGGT GYVPEDGLTA QQLFASADGL TYNDFLILPG FIDFIADEVD LTSALTRKIT LKTPLISSPM DTVTEADMAI AMALMGGIGF IHHNCTPEFQ ANEVRKVKKF EQGFITDPVV LSPSHTVGDV LEAKMRHGFS GIPITETGTM GSKLVGIVTS RDIDFLAEKD HTTLLSEVMT PRIELVVAPA GVTLKEANEI LQRSKKGKLP IVNDCDELVA IIARTDLKKN RDYPLASKDS QKQLLCGAAV GTREDDKYRL DLLTQAGVDV IVLDSSQGNS VYQIAMVHYI KQKYPHLQVI GGNVVTAAQA KNLIDAGVDG LRVGMGCGSI CITQEVMACG RPQGTAVYKV AEYARRFGVP IIADGGIQTV GHVVKALALG ASTVMMGSLL AATTEAPGEY FFSDGVRLKK YRGMGSLDAM EKSSSSQKRY FSEGDKVKIA QGVSGSIQDK GSIQKFVPYL IAGIQHGCQD IGARSLSVLR SMMYSGELKF EKRTMSAQIE GGVHGLHSYE KRLY | |
Greater than 95.0% as determined by SDS-PAGE. |