missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human IL-36 alpha/IL-1F6 (aa 6-158) Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
478.00€
Specifications
Gene ID (Entrez) | 27179 |
---|---|
Molecular Weight (g/mol) | 17 kDa |
Name | IL-36 alpha/IL-1F6 (aa 6-158) |
Quantity | 10 μg |
Source | E. coli-derived human IL-36 alpha/IL-1F6 protein Lys6-Phe158 |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18156950
|
R&D Systems™
6995-IL-010 |
Lyophilized from a 0.2 μm filtered solution in Tris, NaCl, TCEP, EDTA and Tween™ 20 with BSA as a carrier protein. |
478.00€
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18147180
|
R&D Systems™
6995-IL-010/CF |
Lyophilized from a 0.2 μm filtered solution in Tris, NaCl, TCEP, EDTA and Tween™ 20 with Trehalose. |
478.00€
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Recombinant Human IL-36 alpha/IL-1F6 (aa 6-158) Protein is derived from E. coli. The Recombinant Human IL-36 alpha/IL-1F6 (aa 6-158) Protein has been validated for the following applications: Bioactivity.Specifications
27179 | |
IL-36 alpha/IL-1F6 (aa 6-158) | |
E. coli-derived human IL-36 alpha/IL-1F6 protein Lys6-Phe158 | |
Monomer | |
Recombinant | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
17 kDa | |
10 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
Reconstitute at 100μg/mL in Water. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title