Learn More
R&D Systems™ Recombinant Human IL-18/IL-1F4 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand:  R&D Systems™ 9124-IL-500/CF
1453.10 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
 View more versions of this product
Product Code. 18745939
  • 1900.00€ / 500µg
Explore more special offers
Description
Description
The Recombinant Human IL-18/IL-1F4 Protein is derived from E. coli. The Recombinant Human IL-18/IL-1F4 Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
Q14116
3606
500 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Iboctadekin, IFN-gamma-inducing factor, IGIF, IGIFIL-1 gamma, IL18, IL-18MGC12320, IL-1F4, IL1F4iboctadekin, IL-1g, Interferon gamma-inducing factor, interleukin 18 (interferon-gamma-inducing factor), Interleukin-1 gamma, interleukin-18
IL18
Unconjugated
Recombinant
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Bioactivity
Observed M.W.: 19kDa, reducing conditions, Theoretical M.W.: 18kDa
E. coli-derived human IL-18/IL-1F4 protein Tyr37-Asp193
<0.10 EU per 1μg of the protein by the LAL method.
IL-18/IL-1F4
Measured by its ability to induce IFN-gamma secretion by KG-1 human acute myelogenous leukemia cells in the presence of TNF-alpha . The ED50 for this effect is typically 1.5-9ng/mL.
Human
E. coli-derived Tyr37-Asp193