missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GMP reductase 2 Protein
A DNA sequence encoding the Homo sapiens (Human) GMP reductase 2, was expressed in the hosts and tags indicated.
160.00€ - 2385.00€
Specifications
Accession Number | NP_001002000.1 |
---|---|
Gene ID (Entrez) | 51292 |
Name | GMP reductase 2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16012811
|
enQuireBio™
QP8126-ec-10ug |
10 μg | |||||||
16052811
|
enQuireBio™
QP8126-ec-50ug |
50 μg | |||||||
16002811
|
enQuireBio™
QP8126-ec-100ug |
100 μg |
530.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16032811
|
enQuireBio™
QP8126-ec-200ug |
200 μg | |||||||
16042811
|
enQuireBio™
QP8126-ec-500ug |
500 μg | |||||||
16022811
|
enQuireBio™
QP8126-ec-1mg |
1 mg |
2385.00€
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_001002000.1 | |
GMP reductase 2 Protein | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
51292 | |
Research Use Only | |
GMPR2 | |
Recombinant Protein | |
E. coli | |
MTSCLPALRF IATPRLSAMP HIDNDVKLDF KDVLLRPKRS TLKSRSEVDL TRSFSFRNSK QTYSGVPIIA ANMDTVGTFE MAKVLCKFSL FTAVHKHYSL VQWQEFAGQN PDCLEHLAAS SGTGSSDFEQ LEQILEAIPQ VKYICLDVAN GYSEHFVEFV KDVRKRFPQH TIMAGNVVTG EMVEELILSG ADIIKVGIGP GSVCTTRKKT GVGYPQLSAV MECADAAHGL KGHIISDGGC SCPGDVAKAF GAGADFVMLG GMLAGHSESG GELIERDGKK YKLFYGMSSE MAMKKYAGGV AEYRASEGKT VEVPFKGDVE HTIRDILGGI RSTCTYVGAA KLKELSRRTT FIRVTQQVNP IFSEAC | |
Greater than 90% as determined by SDS-PAGE. |