Learn More
R&D Systems™ Recombinant Human Galectin-2 (HEK-293-expressed) Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand: R&D Systems™ 9874-GA-050
285.09 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
- Derived from HEK293
- Validated for the following applications: Bioactivity.
Specifications
P05162 | |
3957 | |
50 μg | |
12 months from date of receipt, -20 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution 3 months, -20 °C under sterile conditions after reconstitution. | |
Unconjugated | |
Reconstitute at 400 μg/mL in water | |
> 95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining |
Bioactivity | |
12 to 15 kDa, reducing conditions | |
Human embryonic kidney cell, HEK293-derived human Galectin-2 protein Met1-Glu132 | |
< 0.10 EU per 1 μg of the protein by the LAL method | |
Human | |
Lyophilized from a 0.2 μm filtered solution in HEPES, NaCl, TCEP, PEG and Trehalose |