missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human FGFR1 beta (IIIc) Fc Chimera Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand: R&D Systems™ 661-FR-050
371.48 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Beschreibung
The Recombinant Human FGF R1 beta (IIIc) Fc Chimera Protein is derived from NS0. The Recombinant Human FGF R1 beta (IIIc) Fc Chimera Protein has been validated for the following applications: Bioactivity.Spezifikation
Lyophilized from a 0.2μm filtered solution in PBS. | |
56 kDa (monomer) | |
50 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
90%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
2260 | |
FGFR1 beta | |
Mouse myeloma cell line,NS0-derived human FGF R1 beta protein, (N-terminus) Human FGF R1 beta (IIIc) (Arg22-Glu285) Accession &Num; NP_075594 IEGRDMD Human IgG1 (Pro100-Lys330) (C-terminus) | |
Disulfide-linked homodimer | |
Recombinant |