missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human EEF1E1 / AIMP3 / p18 Protein

A DNA sequence encoding the Homo sapiens (Human) EEF1E1 / AIMP3 / p18, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5961-ec-50ug

 View more versions of this product

Product Code. 15958959

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_001129122.1
EEF1E1 / AIMP3 / p18 Protein
Research Use Only
EEF1E1
Recombinant Protein
E. coli
AAAAELSLLE KSLGLSKGNK YSAQGERQIP VLQTNNGPSL TGLTTIAAHL VKQANKEYLL GSTAEEKAIV QQWLEYRVTQ VDGHSSKNDI HTLLKDLNSY LEDKVYLTGY NFTLADILLY YGLHRFIVDL TVQEKEKYLN VSRWFCHIQH YPGIRQHLSS VVFIKNRLYT NSH
Greater than 90% as determined by SDS-PAGE.
9521
50 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions