missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Cystathionine beta-synthase Protein

A DNA sequence encoding the Homo sapiens (Human) Cystathionine beta-synthase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5768-ye-100ug

 View more versions of this product

Product Code. 15917629

  • 545.00€ / 100µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_000062.1
Cystathionine beta-synthase Protein
Research Use Only
CBS
Recombinant Protein
Yeast
MPSETPQAEV GPTGCPHRSG PHSAKGSLEK GSPEDKEAKE PLWIRPDAPS RCTWQLGRPA SESPHHHTAP AKSPKILPDI LKKIGDTPMV RINKIGKKFG LKCELLAKCE FFNAGGSVKD RISLRMIEDA ERDGTLKPGD TIIEPTSGNT GIGLALAAAV RGYRCIIVMP EKMSSEKVDV LRALGAEIVR TPTNARFDSP ESHVGVAWRL KNEIPNSHIL DQYRNASNPL AHYDTTADEI LQQCDGKLDM LVASVGTGGT ITGIARKLKE KCPGCRIIGV DPEGSILAEP EELNQTEQTT YEVEGIGYDF IPTVLDRTVV DKWFKSNDEE AFTFARMLIA QEGLLCGGSA GSTVAVAVKA AQELQEGQRC VVILPDSVRN YMTKFLSDRW MLQKGFLKEE DLTEKKPWWW HLR
Greater than 90% as determined by SDS-PAGE.
102724560
100 μg
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions