missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Cyclic GMP-AMP synthase Protein

A DNA sequence encoding the Homo sapiens (Human) Cyclic GMP-AMP synthase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7853-ec-50ug

 View more versions of this product

Product Code. 16051191

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_612450.2
Cyclic GMP-AMP synthase Protein
Research Use Only
MB21D1
Recombinant Protein
E. coli
GASKLRAVLE KLKLSRDDIS TAAGMVKGVV DHLLLRLKCD SAFRGVGLLN TGSYYEHVKI SAPNEFDVMF KLEVPRIQLE EYSNTRAYYF VKFKRNPKEN PLSQFLEGEI LSASKMLSKF RKIIKEEIND IKDTDVIMKR KRGGSPAVTL LISEKISVDI TLALESKSSW PASTQEGLRI QNWLSAKVRK QLRLKPFYLV PKHAKEGNGF QEETWRLSFS HIEKEILNNH GKSKTCCENK EEKCCRKDCL KLMKYLLEQL KERFKDKKHL DKFSSYHVKT AFFHVCTQNP QDSQWDRKDL GLCFDNCVTY FLQCLRTEKL ENYFIPEFNL FSSNLIDKRS KEFLTKQIEY ERNNEFPVFD EF
Greater than 90% as determined by SDS-PAGE.
115004
50 μg
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions