missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CDC42 Protein
A cDNA sequence encoding the CDC42 was constructed and used to recombinantly synthesize the protein.
137.00€ - 2725.00€
Specifications
Name | CDC42 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15907038
|
enQuireBio™
QP11365-10ug |
10 μg |
137.00€
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15927038
|
enQuireBio™
QP11365-50ug |
50 μg |
212.00€
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15917038
|
enQuireBio™
QP11365-1mg |
1 mg |
2725.00€
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
CDC42 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
The CDC42 solution contains 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 10% glycerol and 2mM EDTA. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MASMTGGQQM GRGSHMQTIK CVVVGDGAVG KTCLLISYTT NKFPSEYVPT VFDNYAVTVM IGGEPYTLGL FDTAGQEDYD RLRPLSYPQT DVFLVCFSVV SPSSFENVKE KWVPEITHHC PKTPFLLVGT QIDLRDDPST IEKLAKNKQK PITPETAEKL ARDLKAVKYV ECSALTQKGL KNVFDEAILA ALEPPEPKKS RRC | |
Greater than 95.0% as determined by SDS-PAGE. |