missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BPNT1 Protein
A cDNA sequence encoding the BPNT1 was constructed and used to recombinantly synthesize the protein.
137.00€ - 7875.00€
Specifications
Name | BPNT1 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15936458
|
enQuireBio™
QP11199-2ug |
2 μg |
137.00€
2µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15916458
|
enQuireBio™
QP11199-10ug |
10 μg |
212.00€
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15926458
|
enQuireBio™
QP11199-1mg |
1 mg |
7875.00€
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
BPNT1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The BPNT1 protein solution (0.5 mg/1ml) is formulated in 20mM Tris-HCl buffer (pH 8.0) 5mM DTT, 0.1M NaCl and 10% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSMASS NTVLMRLVAS AYSIAQKAGM IVRRVIAEGD LGIVEKTCAT DLQTKADRLA QMSICSSLAR KFPKLTIIGE EDLPSEEVDQ ELIEDSQWEE ILKQPCPSQY SAIKEEDLVV WVDPLDGTKE YTEGLLDNVT VLIGIAYEGK AIAGVINQPY YNYEAGPDAV LGRTIWGVLG LGAFGFQLKE VPAGKHIITT TRSHSNKLVT DCVAAMNPDA VLRVGGAGNK IIQLIEGKAS AYVFASPGCK KWDTCAPEVI LHAVGGKLTD IHGNVLQYHK DVKHMNSAGV LATLRNYDYY ASRVPESIKN ALVP | |
Greater than 90% as determined by SDS-PAGE. |