missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human ADAM9 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity
Brand: R&D Systems™ 939-AD-020
This item is not returnable.
View return policy
Description
The Recombinant Human ADAM9 Protein is derived from NS0. The Recombinant Human ADAM9 Protein has been validated for the following applications: Enzyme Activity.![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Supplied as a 0.2μm filtered solution in MES and NaCl. | |
55 kDa | |
20 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening. | |
Unconjugated | |
90%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
8754 | |
ADAM9 | |
Mouse myeloma cell line,NS0-derived human ADAM9 protein Ala206-Asp697,with a C-terminal 10-His tag | |
Mature form. Recombinant Human ADAM9 is prone to proteolytic cleavage at C-terminus. The poly-His tag may not be present in the preparation. | |
Recombinant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction