missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ACTR3 Protein

A cDNA sequence encoding the ACTR3 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10954-1mg

 View more versions of this product

Product Code. 15925678

  • 4090.00€ / 1mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

ACTR3 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMAGRLPA CVVDCGTGYT KLGYAGNTEP QFIIPSCIAI KESAKVGDQA QRRVMKGVDD LDFFIGDEAI EKPTYATKWP IRHGIVEDWD LMERFMEQVI FKYLRAEPED HYFLLTEPPL NTPENREYTA EIMFESFNVP GLYIAVQAVL ALAASWTSRQ VGERTLTGTV IDSGDGVTHV IPVAEGYVIG SCIKHIPIAG RDITYFIQQL LRDREVGIPP EQSLETAKAV KERYSYVCPD LVKEFNKYDT DGSKWIKQYT GINAISKKEF SIDVGYERFL GPEIFFHPEF ANPDFTQPIS EVVDEVIQNC PIDVRRPLYK NIVLSGGSTM FRDFGRRLQR DLKRTVDARL KLSEELSGGR LKPKPIDVQV ITHHMQRYAV WFGGSMLAST PEFYQVCHTK KDYEEIGPSI CRHNPVFGVM S
Greater than 85.0% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
ACTR3 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0) and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions