missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Activin RIA/ALK-2 Fc Chimera Protein
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity
Brand: R&D Systems™ 637-AR-100
This item is not returnable.
View return policy
Description
The Recombinant Human Activin RIA/ALK-2 Fc Chimera Protein is derived from NS0. The Recombinant Human Activin RIA/ALK-2 Fc Chimera Protein has been validated for the following applications: Binding Activity.Specifications
Lyophilized from a 0.2μm filtered solution in PBS. | |
Predicted MW: 39kDa & 28kDa (monomers) Observed MW: 40-45kDa & 34-38kDa, reducing conditions | |
100 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
90 | |
Activin RIA/ALK-2 Fc Chimera | |
Mouse myeloma cell line,NS0-derived human Activin RIA/ALK-2 protein, (N-terminus) Human Activin RIA (Asp23-Val124 & Phe120-Val124) Accession &Num; Q53SV1 IEGRMD Human IgG1 (Pro100-Lys330) 6-His tag (C-terminus) | |
Disulfide-linked homodimer | |
Recombinant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction