missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Cynomolgus Monkey TIGIT Fc Chimera Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Brand:  R&D Systems™ 9380-TG-050

414.68 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.


Product Code. 18715017

  • 526.00€ / 50µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Measured by its binding ability in a functional ELISA. When Recombinant Human CD155/PVR (Catalog # 2530-CD ) is coated at 2.5 μg/mL (100 μL/well), the concentration of Recombinant Cynomolgus Monkey TIGIT Fc Chimera that produces 50% optimal binding response is 12-72 ng/mL.

Order Info

Ships at ambient temperature.

Specifications

Specifications

XP_005548158
Lyophilized from a 0.2 μm filtered solution in PBS.
M.W. (Observed): 43 - 60 kDa, reducing conditions; M.W. (theoretical): 40 kDa
Human embryonic kidney cell, HEK293-derived cynomolgus monkey TIGIT protein Cynomolgus Monkey TIGIT (Met22-Pro142) (N-terminus) Accession # XP_005548158 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
RUO
12 to 72ng/mL
Unconjugated
Recombinant
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Bioactivity
201633
50 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
<0.10 EU / 1 μg of the protein by the LAL method.
Recombinant protein
Cynomolgus Monkey
Reconstitute at 200 μg/mL in PBS.
TIGIT
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

For Research Use Only.