missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Cynomolgus Monkey IL-3R alpha Fc Protein

Brand:  R&D Systems™ 10597-R3-050

380.12 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.


Product Code. 30000794

  • 483.00€ / 50µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Measured by its binding ability in a functional ELISA. When Recombinant Cynomolgus Monkey IL-3R alpha Fc Chimera prootein (Catalog #10597-R3) is represent at 1 μg/mL (100 μg/mL), The concentration of Recombinant Human IL-3 Protein (Catalog # 203-IL ) that produces 50% optimal binding response is found to be 8-40 ng/mL.
Specifications

Specifications

EHH61867.1
Bioactivity
50 μg
CD123, CD123 antigen, hIL3Ra, hIL-3 Ra, IL-3 R alpha, IL-3 receptor subunit alpha, IL3R, IL3R alpha, IL-3 R subunit alpha, IL3RA, IL-3 RA, IL-3 R-alpha, IL3RAY, IL3RX, IL3RY, interleukin 3 receptor, alpha (low affinity), interleukin-3 receptor subunit alpha, MGC34174
Lyophilized from a 0.2 μm filtered solution in PBS.
Unconjugated
3563
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Cynomolgus Monkey
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions