missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Bovine IL-4 Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

304.00€

Specifications

Gene ID (Entrez) 280824
Molecular Weight (g/mol) 12.7 kDa
Name IL-4
Quantity 25 μg
Source E. coli-derived bovine IL-4 protein His25-Cys135,with an N-terminal Met
View More Specs

Products 2
Product Code Brand Formulation Price Quantity & Availability  
Product Code Brand Formulation Price Quantity & Availability  
18381408
View Documents Promotion Details
R&D Systems™
2469-BL-025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
304.00€
25µg
Please to purchase this item. Need a web account? Register with us today!
18186110
View Documents
R&D Systems™
2469-BL-025/CF
Lyophilized from a 0.2μm filtered solution in PBS.
304.00€
25µg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The Recombinant Bovine IL-4 Protein is derived from E. coli. The Recombinant Bovine IL-4 Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

280824
IL-4
E. coli-derived bovine IL-4 protein His25-Cys135,with an N-terminal Met
B cell growth factor 1, BCDF, B-cell stimulatory factor 1, BCGF1, BCGF-1, binetrakin, BSF1, BSF-1, IL4, IL-4B_cell stimulatory factor 1, IL4E12, interleukin 4, interleukin-4, Lymphocyte stimulatory factor 1, MGC79402, Pitrakinra
Recombinant
12.7 kDa
25 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Videos
SDS
Documents

Documents

Product Certifications