missing translation for 'onlineSavingsMsg'
Learn More

Heidolph™ Reax Overhead Shakers for Bottles

Product Code. p-7083456 Shop All Heidolph Instruments Products
Click to view available options
Model:
Reax 20/12
Reax 20/8
Holds:
12 Bottles
8 Bottles
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10633201

Brand: Heidolph™ 5412000800

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Easy and fast attachment of bottles. Heidolph Reax Overhead Shakers for Bottles Comes with attachment for 2L bottles.

  • Speed continuously adjustable from 1 to 16rpm
  • Also speed ranges of 0.5 - 8rpm and 2 - 32rpm available
  • Analog dial controls for rpm adjustment
  • Easy and fast attachment of bottles
  • Comes with attachment for 2L bottles
  • Attachment for bottle sizes of 0.5L and 1L available
  • Other vessels of 270mm height and max diameter of 136mm can be fixed with tension plate
  • Protection class IP 21
TRUSTED_SUSTAINABILITY

Specifications

Voltage 100/115/220/230 V
Holds 8 Bottles
Product Type Overhead Shaker
Model Reax 20/8
No. of Positions 8
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.