Learn More
Lenz Laborglasintrumente Reaction Vessels Cylindrical with Thermostatic and Vacuum Jacket and Withdrawal Valve
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
2565.00€ - 6120.00€
Product Code | Brand | Height (Metric) | Capacity (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Height (Metric) | Capacity (Metric) | Price | Quantity & Availability | ||||
11903771
|
Lenz Laborglasintrumente
06061473 |
415 mm | 2 L |
3860.00€
Each |
Estimated Shipment: 21-02-2025
Log in to see stock. |
||||
11913771
|
Lenz Laborglasintrumente
06061479 |
525 mm | 4 L |
6115.00€
Each |
Estimated Shipment: 21-02-2025
Log in to see stock. |
||||
11923761
|
Lenz Laborglasintrumente
06061249 |
315 mm | 0.25 L |
2565.00€
Each |
Estimated Shipment: 21-02-2025
Log in to see stock. |
||||
11923771
|
Lenz Laborglasintrumente
06061482 |
570 mm | 5 L |
6120.00€
Each |
Estimated Shipment: 21-02-2025
Log in to see stock. |
||||
11933761
|
Lenz Laborglasintrumente
06061258 |
330 mm | 0.5 L |
2685.00€
Each |
Estimated Shipment: 21-02-2025
Log in to see stock. |
||||
11953761
|
Lenz Laborglasintrumente
06061270 |
395 mm | 1 L |
2790.00€
Each |
Estimated Shipment: 21-02-2025
Log in to see stock. |
||||
11963761
|
Lenz Laborglasintrumente
06061273 |
490 mm | 2 L |
2875.00€
Each |
Estimated Shipment: 21-02-2025
Log in to see stock. |
||||
11973761
|
Lenz Laborglasintrumente
06061276 |
310 mm | 3 L |
3240.00€
Each |
Estimated Shipment: 21-02-2025
Log in to see stock. |
||||
11983761
|
Lenz Laborglasintrumente
06061279 |
460 mm | 4 L |
3350.00€
Each |
Estimated Shipment: 21-02-2025
Log in to see stock. |
||||
11993761
|
Lenz Laborglasintrumente
06061476 |
460 mm | 3 L |
3685.00€
Each |
Estimated Shipment: 21-02-2025
Log in to see stock. |
||||
Description
- With graduation
- With laboratory flange LF, with groove
- With two connectors (with ball flanges KF 15 /KF 25)
- Made of DURAN™ tubing
- Other sizes on request
Specifications
Duran Glass |