missing translation for 'onlineSavingsMsg'
Learn More

Rat μ Opioid R/OPRM1 Antibody, R&D Systems™

Rabbit Monoclonal Antibody

189.00€ - 388.00€

Specifications

Antigen mu Opioid R/OPRM1
Clone 1126D
Dilution Western Blot 0.5 ug/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18723698
View Documents
R&D Systems
MAB86291-SP
25 μg
189.00€
25µL
Please to purchase this item. Need a web account? Register with us today!
18778039
View Documents
R&D Systems
MAB86291
100 μg
388.00€
100µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

mu Opioid R/OPRM1 Monoclonal specifically detects mu Opioid R/OPRM1 in Rat samples. It is validated for Western Blot.
Specifications

Specifications

mu Opioid R/OPRM1
Western Blot 0.5 ug/mL
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide
hMOP, KIAA0403, LMOR, MOP, mOpioidR, MOR, MOR1, M-OR-1, MOR-1, MOR1MOR, Mu opiate receptor, Mu opioid receptor, mu-type opioid receptor, opioid receptor, mu 1, OPRM, OPRM1
OPRM1
IgG
Protein A or G purified from cell culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
1126D
Western Blot
Unconjugated
Rabbit
Rat
P33535
4988
Synthetic peptide with a short amino acid sequence from the N-terminus of rat μ Opioid R/OPRM1 Accession # P33535
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects rat μ Opioid R/OPRM1 in direct ELISAs.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only