missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat μ Opioid R/OPRM1 Antibody, R&D Systems™
Rabbit Monoclonal Antibody
Brand: R&D Systems MAB8629-SP
This item is not returnable.
View return policy
Description
mu Opioid R/OPRM1 Monoclonal specifically detects mu Opioid R/OPRM1 in Rat samples. It is validated for Immunohistochemistry.Specifications
mu Opioid R/OPRM1 | |
Monoclonal | |
Unconjugated | |
Supplied as a solution in PBS containing BSA, Glycerol and Sodium Azide. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide | |
hMOP, KIAA0403, LMOR, MOP, mOpioidR, MOR, MOR1, M-OR-1, MOR-1, MOR1MOR, Mu opiate receptor, Mu opioid receptor, mu-type opioid receptor, opioid receptor, mu 1, OPRM, OPRM1 | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
RUO | |
4988 | |
Rat | |
Purified |
Immunohistochemistry | |
1126B | |
Immunohistochemistry 1:125 dilution | |
P33535 | |
OPRM1 | |
Synthetic peptide with a short amino acid sequence from the N-terminus of rat μ Opioid R/OPRM1 Accession # P33535 | |
25 μg | |
Primary | |
Detects rat μ Opioid R/OPRM1 in direct ELISAs. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG |
For Research Use Only