missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat LYVE-1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF7939-SP
This item is not returnable.
View return policy
Description
LYVE-1 Polyclonal specifically detects LYVE-1 in Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| LYVE-1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| cell surface retention sequence binding protein-1, Cell surface retention sequence-binding protein 1, CRSBP1, CRSBP-1, extracellular link domain containing 1, extracellular link domain-containing 1, Extracellular link domain-containing protein 1, HAR, Hyaluronic acid receptor, lymphatic vessel endothelial hyaluronan receptor 1, lymphatic vessel endothelial hyaluronic acid receptor 1, LYVE1, LYVE-1XLKD1, XLKD1 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 10894 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| NP_001099756 | |
| LYVE1 | |
| Mouse myeloma cell line NS0-derived recombinant rat LYVE-1 Thr53-Thr259 Accession # NP_001099756 | |
| 25 μg | |
| Primary | |
| Detects rat LYVE-1 in direct ELISA and Western blots. In direct ELISAs, approximately 50% cross-reactivity with recombinant mouse LYVE-1 is observed, and approximately 10% cross-reactivity with recombinant human LYVE-1 is observed. | |
| Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction