Learn More
OHAUS™ Ranger™ Count 4000
Product Code. 15513903 Shop All Ohaus Products
Click to view available options
Capacity:
3.00 kg
6.00 kg
15.00 kg
30.00 kg
Model:
RC41M15
RC41M15-M
RC41M3
RC41M3-M
RC41M30-M
RC41M6-M
Unit Size:
Each
Regulatory: This product is under eco-contribution tax.
Product Code. 15513903
Brand: OHAUS™ 30237023
Regulatory: This product is under eco-contribution tax.
OHAUS™ Ranger™ Count 4000 is ideal for tough industrial counting jobs that require durability and accuracy. This scale is equipped with weighing, parts counting, check counting, and accumulation functions
  • Sturdy metal housing and slip-resistant rubber feet provide the protection, stability and long product life needed for tough industrial weighing.
  • Accurate weighing and counting performance is assured with 1:1,500,000 internal counting resolution and auto-optimization software that reduces errors.
  • Easy-to-operate with SmarText™ software, a centrally located count display and dedicated Sample and Tare keys.
TRUSTED_SUSTAINABILITY
Specifications
Battery Life 210 Hours with Rechargeable Battery
Connection Type Easy access communication port including standard RS232 (included) and second RS232, USB or Ethernet (accessories sold separately)
Construction Rugged Cast Aluminum Housing, Stainless Steel Platform
Capacity 30.00 kg
Display Type 3 Window Backlit LCD, 3 Color Checkweighing LEDs
Readability 10.000000 g
Linearity 2.000000 g
Model RC41M30-M
Repeatability 2.000000 g
Temperature (Metric) Operating -10°C to +40°C
Weighing Modes Gram, Kilogram
Dimensions (L x W x H) 327 x 315 x 122 mm
Weight (Metric) 7.000 kg