Learn More
Ohaus™ Ranger™ Count 3000 Multipurpose Compact Counting Scale
Compact, multipurpose counting scale with weighing, parts counting, check counting, and accumulation functions.
Brand:  Ohaus™ 30060907
 View more versions of this product
Product Code. 15853595
  • 631.00€ / Each
Explore more special offers
Description
Description
  • Precise counting and weighing performance with 30 item library
  • The most user-friendly counting scale on the market
  • Rapid stabilization provides unmatched throughput
TRUSTED_SUSTAINABILITY
Specifications
Specifications
210 Hours with Rechargeable Battery
ABS housing, stainless steel platform, numeric keypad, menu lock switch, up front level indicator, transportation handle
3-window Backlight LCD + 3 LED
RS232, AC power, rechargeable battery
Stainless Steel Platform
0.100000 g
311.00 x 327.00 x 117.000 mm
311.00 mm
12.90 in.
300 mm
117.000 mm
311.00 mm
Ethernet (Available as an Accessory)USB (Available as an Accessory)RS232 (Included)2nd RS232 (Available as an Accessory)
1.5 kg
0.500000 g
0.100000 g
RC31P1502-M
311.00 x 327.00 mm
12.20 in.
225 mm
327.00 mm
4.60 in.
12.20 in.
100/240 VAC 50/60 Hz internal universal power supply, internal rechargeable lead acid battery