missing translation for 'onlineSavingsMsg'
Learn More

rac BHFF, Tocris Bioscience™

Product Code. 18014762
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent and selective postitive allosteric modulator of GABAB receptors

Potent and selective GABAB receptor positive allosteric modulator that increases the potency and efficacy of GABA ( > 15-fold and > 149% respectively). Exhibits anxiolytic activity in vivo and is orally active.

Chemical Identifiers

CAS 123557-91-5
Molecular Formula C17H21F3O3
Molecular Weight (g/mol) 330.347
InChI Key RVNOANDLZIIFHB-UHFFFAOYSA-N
Synonym rac bhff, bhff, 2 3h-benzofuranone,5,7-bis 1,1-dimethylethyl-3-hydroxy-3-trifluoromethyl, acmc-20emwz, rac bhff hplc, 5,7-ditert-butyl-3-hydroxy-3-trifluoromethyl-1-benzofuran-2-one, 3h-benzofuran-2-one, 5,7-di-tert-butyl-3-hydroxy-3-trifluoromethyl, 5,7-di tert-butyl-3-hydroxy-3-trifluoromethyl-1-benzofuran-2 3h-one, 5,7-di-tert-butyl-3-hydroxy-3-trifluoromethyl-1-benzofuran-2 3h-one, 5,7-bis 1,1-dimethylethyl-3-hydroxy-3 trifluoromethyl-2 3h-benzofuranone
PubChem CID 4332683
IUPAC Name 5,7-ditert-butyl-3-hydroxy-3-(trifluoromethyl)-1-benzofuran-2-one
SMILES CC(C)(C)C1=CC(=C2C(=C1)C(C(=O)O2)(C(F)(F)F)O)C(C)(C)C

Specifications

Quantity 50 mg
Formula Weight 330.34
Percent Purity >98%
Chemical Name or Material rac BHFF
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'