missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Rabbit IFN-gamma ELISA Kit
Rabbit IFN-gamma quantitates rabbit IFN-gamma in serum, plasma, supernatant.
690.00€ - 2690.00€
Specifications
Accession Number | BoIFNG; cytokine; EGK_03901; gamma interferon; gamma interferon precursor; gamma-interferon; gamma-interferon precursor (AA -23 to 143); H-IFN-g; Ifg; IFI; IFN gamma; IFN ?; IFNG; IFN-g; IFNG2; IFN-gamma; IFN-gamma precursor; IFN-y; IFN?; Immune interferon; interferon gamma; interferon gamma precursor; interferon gamma type 2; Interferon y; Interferon ?; interferon, gamma; interferon-gama; interferon-gamma; Interferon-gamma level; interferon-gamma precursor; Interferon?; M-IFN-g; R-IFN-g |
---|---|
Assay Range | 0.16 to 100 ng/mL |
Assay Sensitivity | 0.16 ng/mL |
Conjugate | Biotin |
Product Type | ELISA |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17155839
|
Invitrogen™
ER4RBX5 |
5 x 96 Tests |
2690.00€
Each |
Estimated Shipment: 05-02-2025
Log in to see stock. |
|||||
17175013
|
Invitrogen™
ER4RB |
96 Tests |
690.00€
Pack of 96 |
Estimated Shipment: 06-02-2025
Log in to see stock. |
|||||
Description
Rabbit IFN-gamma quantitates rabbit IFN-gamma in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant rabbit IFN-gamma.
IFN gamma (Interferon gamma, Type II interferon) is a macrophage activation factor, and immune interferon that is produced primarily by T-lymphocytes and natural killer cells in response to antigens, mitogens, Staphylococcus enterotoxin B, phytohemaglutanin and other cytokines. IFN gamma is a dimeric protein consisting of two 146 amino acid subunits. IFN gamma is a glycoprotein that exists, functionally, as a homodimer of approximately 45 kDa. On SDS-PAGE, IFN gamma appears as a combination of 25, 20 and minor 15.5 kDa bands as a result of differential glycosylation. The biological activity of the IFN gamma homodimer is highly species specific. Human IFN gamma does not show cross-reactivity with mouse. IFN gamma function includes the following: antiviral activity, tumor antiproliferative activity, induction of class I and II MHC, macrophage activation, and enhanced immunoglobulin secretion by B lymphocytes. IFN gamma is involved in cytokine regulation and also acts synergistically with other cytokines. Activation of IFN gamma takes place through binding of IFN gamma receptor I and II, and activating the JAK-STAT pathway. IFN gamma does not show any homology with IFN alpha or IFN beta but human IFN gamma shows about 40% sequence homology with mouse IFN gamma. IFN gamma is upregulated by IL2, FGF basic, EGF and downregulated by vitamin D3 or DMN. IFN gamma gene mutations are associated with aplastic anemia.Specifications
BoIFNG; cytokine; EGK_03901; gamma interferon; gamma interferon precursor; gamma-interferon; gamma-interferon precursor (AA -23 to 143); H-IFN-g; Ifg; IFI; IFN gamma; IFN ?; IFNG; IFN-g; IFNG2; IFN-gamma; IFN-gamma precursor; IFN-y; IFN?; Immune interferon; interferon gamma; interferon gamma precursor; interferon gamma type 2; Interferon y; Interferon ?; interferon, gamma; interferon-gama; interferon-gamma; Interferon-gamma level; interferon-gamma precursor; Interferon?; M-IFN-g; R-IFN-g | |
0.16 ng/mL | |
ELISA | |
Colorimetric Microplate Reader | |
IFNG | |
<10% | |
HRP | |
Plasma, 50 μL; Supernatant, 50 μL; Serum, 50 μL | |
IFN gamma | |
4 hr 45 min |
0.16 to 100 ng/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
P30123 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
Research Use Only | |
2°C to 8°C | |
1 hr 20 min |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title