Learn More
LAT2 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant LAT2.
Brand: Abnova PAB20215.100uL
Description
This gene is one of the contiguous genes at 7q11.23 commonly deleted in Williams syndrome, a multisystem developmental disorder. This gene consists of at least 14 exons, and its alternative splicing generates 3 transcript variants, all encoding the same protein. [provided by RefSeq
Sequence: AKRSEKIYQQRSLREDQQSFTGSRTYSLVGQAWPGPLADMAPTRKDKLLQFYPSLEDPASSRYQNFSKGSRHGSEEAYIDPIAMEYYNWGRFSKPPEDDDANSYENVLICKQKTTETGAQQEGIGGLCRGDLSLSLALKTGPTSpecifications
| LAT2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant LAT2. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| HSPC046/LAB/NTAL/WBSCR15/WBSCR5/WSCR5 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 7462 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
| LAT2 | |
| LAT2 | |
| Recombinant protein corresponding to amino acids of human LAT2. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only