missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Bovine IgG (H+L) Secondary Antibody, TRITC, Invitrogen™
Rabbit Polyclonal Secondary Antibody
Brand: Invitrogen PA128691
This item is not returnable.
View return policy
Description
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Anti-Bovine secondary antibodies are affinity-purified antibodies with well-characterized specificity for bovine (cow, ox) immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Specifications
Bovine IgG (H+L) | |
Polyclonal | |
TRITC | |
Rabbit | |
IgG fraction | |
RUO | |
Bovine | |
Liquid |
Flow Cytometry, Immunocytochemistry | |
1 mg/mL | |
0.02M potassium phosphate with 0.15M NaCl, 10mg/mL BSA and 0.01% sodium azide; pH 7.2 | |
Full length native bovine IgG (purified). | |
1 mg | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction