Learn More
RAB4A Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10347-1-AP-20UL
Description
The RAB4A antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human RAB4A. This antibody recognizes human, mouse, rat antigen. The RAB4A antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
| RAB4A | |
| Polyclonal | |
| Unconjugated | |
| 20 μL | |
| 5867 | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| 200 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Rat, Mouse |