Learn More
R&D Systems™ Human Common gamma Chain (NP_000197) VersaClone cDNA
CD132; IL-2 R gamma; IL2RG
Brand:  R&D Systems™ RDC1129
Product Code. 18700539
  • 322.00€ / 10µg
Explore more special offers
Description
Description
The Human Common gamma Chain (NP_000197) VersaClone cDNA clone contains the complete ORF for Common gamma Chain/IL-2 R gamma, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The Common gamma Chain/IL-2 R gamma insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature
Specifications
Specifications
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
For Reasearch Use Only; Not for Use in Humans