Learn More
R&D Systems™ Mouse Prokineticin R2/PROKR2 (NP_659193) VersaClone cDNA
Prokineticin Receptor 2
Brand: R&D Systems™ RDC0674
This item is not returnable.
View return policy
Description
The Mouse Prokineticin R2/PROKR2 (NP_659193) VersaClone cDNA clone contains the complete ORF for Prokineticin R2/PROKR2, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The Prokineticin R2/PROKR2 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.Order Info
Ships at ambient temperature.
Specifications
mPROKR2 | |
10 μg at 0.2 μg/μL | |
1157bp | |
Mouse |
NP_659193 | |
cDNA is provided in 10mM Tris-Cl, pH 8.5 | |
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. | |
10 μg |
Product Content Correction
Product Title
For Reasearch Use Only; Not for Use in Humans