missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse beta-3 Adrenergic R/ADRB3 (NP_038490) VersaClone cDNA
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Description
The Mouse beta-3 Adrenergic R/ADRB3 (NP_038490) VersaClone cDNA clone contains the complete ORF for beta-3 Adrenergic R/ADRB3, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The beta-3 Adrenergic R/ADRB3 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature.
Specifications
Specifications
| Gene | mADRB3 |
| Accession Number | NP_038490 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10mM Tris-Cl, pH 8.5 |
| Insert Type | 1215bp |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Mouse |
| Quantity | 10 μg |
For Reasearch Use Only; Not for Use in Humans
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction