missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Syndecan-1 Protein, CF
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand: R&D Systems™ 2780-SD-050
This item is not returnable.
View return policy
Description
Measured by the ability of the immobilized protein to enhance the adhesion of Saos-2 human osteosarcoma cells to human Fibronectin. Measured by the ability of the immobilized protein to enhance the adhesion of Saos-2 human osteosarcoma cells to human Fibronectin (0.5 μg/mL, Catalog # 4305-FNB ). The ED50 for this effect is 2.5-10 μg/mL.Specifications
NP_002988 | |
Lyophilized from a 0.2 μm filtered solution in PBS. | |
M.W. (Observed): 60-90 kDa, reducing conditions; M.W. (theoretical): 24.9 kDa | |
50 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. | |
SDC1 | |
Recombinant | |
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining. |
Bioactivity | |
6382 | |
Syndecan-1/CD138 | |
Mouse myeloma cell line, NS0-derived human Syndecan-1/CD138 protein Gln18-Glu251, with a C-terminal 6-His tag | |
<0.10 EU / 1 μg of the protein by the LAL method. | |
Unconjugated | |
Reconstitute at 500 μg/mL in PBS. | |
Syndecan-1/CD138 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction