Learn More
R&D Systems™ Human Complement Component C5aR1 (NP_001727) VersaClone cDNA
C5AR1
Brand:  R&D Systems™ RDC0104
Product Code. 18422610
  • 322.00€ / 10µg
Explore more special offers
Description
Description
The Human Complement Component C5aR1 (NP_001727) VersaClone cDNA clone contains the complete ORF for Complement Component C5a R1, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The Complement Component C5a R1 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature.
Specifications
Specifications
hC5aR
10 μg at 0.2 μg/μL
1066bp
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
10 μg
NP_001727
cDNA is provided in 10mM Tris-Cl, pH 8.5
C5A, C5a anaphylatoxin chemotactic receptor, C5aR, C5a-R, C5AR1, C5ARC5a anaphylatoxin receptor, C5R1C5a ligand, CD88, CD88 antigen, complement component 5 receptor 1, complement component 5 receptor 1 (C5a ligand), complement component 5 A receptor 1, Complement Component C5a R1
Human
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
For Reasearch Use Only; Not for Use in Humans