Learn More
Quickfit™ Vertical Splash Head Adapter
Usually inserted between a flask and condenser to prevent 'carry-over' of raw liquid
Brand: Quickfit SH17/23
58.66 EUR valid until 2024-12-31
Use promo code "23259" to get your promotional price.
Additional Details : Weight : 0.07000kg
Description
Usually inserted between a flask and condenser to prevent carry over of raw liquidSpecifications
Vertical Splash Head Adapter | |
Borosilicate Glass (3.3) | |
24/29 |