Learn More
Quickfit™ Borosilicate Glass Sintered Dip Tube for Dreschel Bottle
Product Code. p-8002652
missing translation for 'orderingAttributeHoverText'
For Use With (Equipment):
For model MF29/23/250 Dreschel bottle
For model MF29/3/125 Dreschel bottle
For model MF29/3/250 Dreschel bottle
For model MF29/3/500 Dreschel bottle
Description:
Stem Length: 132 mm
Stem length: 165 mm
Stem length: 217 mm
Porosity:
0
1
2
3
missing translation for 'unitSize'
Each
Pack of 5
Product Code. 10637615
missing translation for 'mfr': Quickfit MF48/13/250
Only null left
Add to basket
Add to basket
Disperse gas into bubbles with QuckFit™ sintered borosilicate glass tubes designed to fit a range of QuickFit™ Deschel bottles. Tubes come in a variety of porosity for fine control over bubble size.
Disperse gas into bubbles with QuckFit™ sintered borosilicate glass tubes designed to fit a range of QuickFit™ Deschel bottles. Tubes come in a variety of porosity for fine control over bubble size.
  • Complies with BS 2461
  • Sintered dip tube disperses gas into bubbles for greater efficiency
  • Choice of sinter porosities to achieve fine bubble size
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance BS 2461
Description Stem length: 165 mm
Material Borosilicate Glass
For Use With (Equipment) For model MF29/3/250 Dreschel bottle
Porosity 1