Learn More
Quickfit™ Borosilicate Glass Erlenmeyer Flask with Quickfit Ground Glass Socket
Click to view available options
Capacity (Metric):
5 mL
25 mL
50 mL
100 mL
150 mL
250 mL
500 mL
1000 mL
2000 mL
Height (Metric):
54 mm
70 mm
85 mm
90 mm
100 mm
116 mm
118 mm
130 mm
175 mm
220 mm
270 mm
Diameter (Metric) Outer:
25 mm
42 mm
51 mm
64 mm
74 mm
85 mm
101 mm
130 mm
160 mm
Socket Joint:
14/23
19/26
24/29
29/32
34/35
Förpackningsstorlek:
Pack of 10
Pack of 2
Pack of 20
Pack of 5
Description
Manufactured from borosilicate glass. Quickfit Jointed Ware™ Quickfit™ Glass Erlenmeyer Flask with Quickfit Ground Glass Socket complies to ISO 4797.
- Complies to ISO 4797
- Conical flasks with highest quality Quickfit™ ground glass socket
- Borosilicate glass for corrosion free performance and high resistance to thermal shock
- Designed for the estimation of arsenic and antimony
- Method uses the formation of a complex between arsine and silver diethyldithiocarbamate with subsequent measurement at 538nm or 5380.
Specifications
Specifications
| Certifications/Compliance | ISO 4797 |
| Capacity (Metric) | 150 mL |
| Material | Borosilicate Glass |
| Height (Metric) | 118 mm |
| Includes | Ground socket |
| Shape | Conical |
| Socket Joint | 19/26 |
| Diameter (Metric) Outer | 74 mm |