Learn More
Quickfit™ Borosilicate Glass Column Packing Support
Designed to support Raschig rings within plain fractionating columns.
92.30€ - 131.00€
Specifications
For Use With (Application) Designed to support Raschig rings within plain fractionating columns
Type Column packing support
Material Pyrex Borosilicate Glass
View More Specs
Products 3
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
15557800
View Documents
Quickfit
EX13/12
19/26 socket size
92.30€
Pack of 5
15567800
View Documents
Quickfit
EX13/25
34/35 socket size
131.00€
Pack of 5
15577800
View Documents
Quickfit
EX13/26
40/38 socket size This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A
Description
Description
  • Support is passed through socket of column and rests at the constriction above the cone
  • Manufactured from chemically resistant borosilicate glass
Specifications
Specifications
Designed to support Raschig rings within plain fractionating columns
Column packing support
Pyrex Borosilicate Glass