Learn More
Applied Biosystems™ QuantStudio™ 6 Pro Real-Time PCR System Extended Warranty Package
Product Code. 17102126 Shop All Applied Biosystems Products
Click to view available options
:
96-well plate
Regulatory: This product is under eco-contribution tax.
Product Code. 17102126
Brand: Applied Biosystems™ A47204
Regulatory: This product is under eco-contribution tax.
Adds innovative smart features to the real-time PCR (qPCR) workflow, all in a compact footprint.
  • Includes facial authentication, voice commands, Smart Help, multiplex capability, motorized interchangeable blocks, simplified workflow, intuitive software, and large capacitive touch-screen interface. The QuantStudio 6 Pro system provides exceptional reproducibility with minimal well-to-well and instrument-to-instrument variation. Designed for ultimate ease-of-use, the system offers high quality, excellent reliability, and an optimal user experience for researchers who want to work smarter in the lab. This package includes the QuantStudio 6 Pro Real-Time PCR System, SmartStart orientation, and an extended warranty.
  • Easy and convenient hands-free operation with facial authentication and voice commands
  • Reduced errors and manual entries with smart qPCR workflow and TaqMan Array Plates with RFID
  • Space savings with small footprint and true standalone operation; set-up, start, monitor, and analyze directly on the touchscreen
  • Application versatility with easily changed blocks and three different block types
  • Increased productivity with automation compatibility for 24/7 operation
  • Access, analyze, and share data anytime, anywhere with our Connect cloud-based platform for real-time information access, secure data storage, and online results sharing
  • Remotely monitor experiments using the Instrument Connect monitoring app
  • Design and Analysis software compatible with PC and Mac operating systems
  • Compatible with over 8 million TaqMan Assays for gene expression, genetic variation, gene regulation, or protein expression experiments
  • Designed for a simple and powerful qPCR experience. Hands-free operation with facial authentication and voice commands enables users to quickly and easily access their personal profiles and settings without even removing their gloves.
  • With embedded RFID readers, the QuantStudio 6 Pro system has the capability to run TaqMan Array Plates with RFID tags. These RFID tags simplify data tracking (with embedded lot number, expiration date, etc.) and reduce the need for manual data entry with preloaded assay layout, dye information, and thermal protocol. Automating this data transfer allows users to simply load the plate and start the run—no data entry needed.
  • Minimize downtime with premium service offerings for the QuantStudio 6 Pro system. Smart Help allows users to contact Technical Support directly from the touchscreen, without the need to download files or spend time on the phone. Experienced technicians will analyze the request and respond according to the user’s preference (phone or email) and resolve the issue as quickly as possible.
  • The Connect platform provides scientists with a secure place to store, analyze, and share data. Applied Biosystems qPCR Analysis Modules within the platform are web browser-based software tools that are flexible, fast, and easy to use. These software tools support a variety of analyses including relative quantitation, standard curve, high resolution melt (HRM), genotyping, and presence/absence. They are compatible with Mac or PC as well as most Applied Biosystems instrument file types.
  • Accommodates the interchange of 96-well (0.2 mL), 96-well (0.1 mL), and 384-well blocks. The system is delivered with one block type; other block types may be purchased separately. I For a system that supports TaqMan array cards and automation, please see our QuantStudio 7 Pro Real-Time PCR System. And for the ultimate in flexibility, consider the QuantStudio 12K Flex Real-Time PCR System, which also accommodates TaqMan OpenArray plates.
  • Simple workflows, reliable assays and reagents, multiple formats, and intuitive software are available for a broad range of applications, including gene expression, genetic variation, and gene regulation.
  • Improved well-to-well and instrument-to-instrument data accuracy is achieved with the OptiFlex system, which features six decoupled excitation and emission filter channels with 21 filter combinations for maximum multiplexing and chemistry flexibility.
  • Detect changes in target quantity as small as 1.5-fold in single-plex reactions and obtain 10 logs of linear dynamic range.
TRUSTED_SUSTAINABILITY
Specifications
Block Format Interchangeable
Format 96-well Plate
For Use With (Equipment) QuantStudio™, ViiA™ 6 System
Capacity 0.1 mL
Product Line QuantStudio
Display Type Touchscreen
Shipping Condition Room Temperature