missing translation for 'onlineSavingsMsg'
Learn More
Learn More
QSOX2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA590300
This item is not returnable.
View return policy
Description
Immunogen sequence: VKKLLEMLQE WLASLPLDRI PYNAVLDLVN NKMRISGIFL TNHIKWVGCQ GSRSELRGYP CSLWKLFHTL TVEASTHPDA LVGTGFEDDP QAVLQTMRRY VHTFFGCKEC GEHFEEMAKE SMDSVKTPDQ AILWLWKKHN MVNGRLAGHL SEDPRFPKLQ WPTPDLCPAC HEEIKGLASW DEGHVLTFLK QHYGRDNLLD TYSADQGDSS EGGTLARGEE EEKRLTPPEV SHGDRDTQSV RPPGALGPRP ALPESLHHSL DGKLQSLDGP GAHKEVGGAA P; Positive Samples: Mouse pancreas, Mouse testis; Cellular Location: Cell membrane, Membrane, Nucleus membrane, Single-pass membrane protein
QSOX2 is a member of the sulfhydryl oxidase/quiescin-6 (Q6) family (QSOX1) that regulates the sensitization of neuroblastoma cells for IFN-gamma (IFNG)-induced cell death.Specifications
QSOX2 | |
Polyclonal | |
Unconjugated | |
Qsox2 | |
BC030934, neuroblastoma-derived sulfhydryl oxidase, QSCN6L1, QSOX2, quiescin Q6 sulfhydryl oxidase 2, quiescin Q6-like 1, quiescin Q6-like protein 1, quiescin sulfhydryl oxidase 2, SOXN, Sulfhydryl oxidase 2, thiol oxidase 2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
227638 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1.95 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal, pH 7.3 | |
Q3TMX7 | |
Qsox2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 370-650 of human QSOX2 (NP_8590523) | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |