missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ qPCR & Go SYBR Hi-ROX Kit

Product Code. p-7139385 Shop All MP Biomedicals Products
Click to view available options
No. of Reactions:
2000 Reactions
500 Reactions
Unit Size:
2000 reactions
500 reactions
This item is not returnable. View return policy

Product Code. 15848541

Brand: MP Biomedicals™ 11EBI01050

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Fast and sensitive qPCR SYBR Master Mix for most ABI platforms.

  • The qPCR & Go SYBR™ Hi-ROX Kit has been developed for fast, highly reproducible real-time PCR and has been validated on commonly used real-time PCR instruments.
  • A combination of the latest advances in buffer chemistry and enhancers, together with an antibody-mediated hot-start DNA polymerase system, ensures that the qPCR & Go SYBR™ Hi-ROX Kit delivers fast, highly-specific and ultra-sensitive real-time PCR.
  • For ease-of-use and added convenience, qPCR & Go SYBR™ Hi-ROX is provided as a 2x mastermix containing all the components necessary for real-time PCR, including the SYBR™Green I dye, dNTPs, stabilisers and enhancers.
  • The kit consists of a ready-to-use premix, only primers and template need to be added.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -20°C
Format Kit
Product Type qPCR Kit
Quantity 5 x 1 mL
For Use With (Application) Real Time PCR (qPCR)
No. of Reactions 500 Reactions

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.