missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyrex™ Borosilicate Glass Reagent Bottles with Glass Stopper
Click to view available options
Capacity (Metric):
50 mL
Specifications
Specifications
Capacity (Metric) | 50 mL |
Includes | Glass stopper |
Material | Borosilicate Glass |
Certifications/Compliance | ISO 3585 |
Closure Included | Yes |
Height (Metric) | 103 mm |
Closure Material | Glass |
Closure Size | 19/26 mm |
Diameter (Metric) | 43 mm |
Closure Type | Stopper |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction