Learn More
Pyrex™ Borosilicate Glass Hollow Blown Stopper
Hollow-blown Quickfit™ jointed glassware featuring C length ground glass sockets.
Brand:  Quickfit 2595/01M
164.35 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 11347804
  • 178.00€ / Pack of 20
Estimated Shipment: 02-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Ideally suited for use with Quickfit™ jointed glassware and other glassware having the shorter 'C' length ground glass sockets
  • All Quickfit™ stoppers conform to the standard tapers as outlined in ISO 383
  • Borosilicate glass for chemical durability and high temperature performance
  • Hollow-blown construction to reduce weight
  • Hexagonal head prevents the stopper from rolling and provides a good grip
  • Top of stopper is flat or slightly concave, allowing them to be stood on end and avoid contamination of the ground surface
Certifications
ISO 383
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ISO 383
Cone size: 10/13
42 mm
Hollow
Borosilicate Glass
Borosilicate Glass
Conical