missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Pyr 10

Selective TRPC3 inhibitor

187.00€ - 682.00€

Specifications

Chemical Name or Material N-[4-[3,5-bis(Trifluoromethyl)-1 H-pyrazol-1-yl]phenyl]-4-methylbenzenesulfonamide
CAS 1315323-00-2
Target TRPC Channel Blockers
Molecular Formula C18H13F6N3O2S
Purity 0.98
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18758536
View Documents Promotion Details
Tocris Bioscience™
6941/10
10 mg
187.00€
10mg
Please to purchase this item. Need a web account? Register with us today!
18768946
View Documents
Tocris Bioscience™
6941/50
50 mg
682.00€
50mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Pyr 10 is a TRPC3 (transient receptor potential canonical 3) channel inhibitor (IC50 = 0.72 μM for TRPC3 mediated calcium entry in vitro), which displays approximately 18-fold selectivity for TRPC3-mediated receptor operated calcium entry (ROCE) over STIM1/Orai1-mediated store operated calcium entry (SOCE). Pyr 10 inhibits NFATc3 activation and inhibits proliferation of rat ventricular cardiac fibroblasts.
Specifications

Specifications

N-[4-[3,5-bis(Trifluoromethyl)-1 H-pyrazol-1-yl]phenyl]-4-methylbenzenesulfonamide
TRPC Channel Blockers
0.98
1315323-00-2
C18H13F6N3O2S
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Product Certifications