Learn More
Euro Protection™ Purofort™ Safety Boots
Euro Protection™ Purofort™ Safety Boots provide flexible and lightweight protection and cold insulation and feature a steel toe cap for protection.
Brand:  Euro Protection™ 9PUSG45
 View more versions of this product
Product Code. 15829236
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Explore more special offers
Description
Description
  • Flexible and lightweight for use all day long
  • Cold insulation
  • Tear and wear resistant
  • Purofort™ upper
  • Polyamide lining
  • Polyurethane insole
  • Steel toe cap
  • Steel, puncture-resistant sole
  • Purofort™ outsole
  • Suitable for use in farming industry
Specifications
Specifications
Green
Polyamide
Polyurethane insole, steel, puncture-resistant sole, Purofort™ outsole.
Steel
EN ISO 20345 : 2011
Agriculture/Farming
45
Purofort™
900 g per boot in size 9.5
Safety Boot