Learn More
Minebea Intec™ Puro™ Floor Scales
Puro™ floor scales are a sturdy weighing solution for diverse industrial weighing needs.
POA
Specifications
Type Floor Scale
Display Type Backlit LCD
Protection Class IP43 (Indicator), IP67 (Platform)
View More Specs
Products 8
Product Code Brand Dimensions (L x W x H) Includes Capacity Readability Price Quantity & Availability  
Product Code Brand Dimensions (L x W x H) Includes Capacity Readability Price Quantity & Availability  
17738949
Encompass_Preferred
View Documents
Minebea Intec™
EF-4PLLP1500-3d
1000 x 1000 x 105 mm Plastic Indicator, Painted Carbon Steel Frame and Load Plate, USB-C Port, 35 mm Weight Display 1500 kg 0.5 kg This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
17748949
Encompass_Preferred
View Documents
Minebea Intec™
EF-4PLLS1500-3d
1000 x 1000 x 105 mm Plastic Indicator, Stainless Steel Frame and Load Plate, USB-C Port, 35 mm Weight Display 1500 kg 0.5 kg This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
17758949
Encompass_Preferred
View Documents
Minebea Intec™
EF-4PNNP1500-3d
1250 x 1250 x 105 mm Plastic Indicator, Painted Carbon Steel Frame and Load Plate, USB-C Port, 35 mm Weight Display 1500 kg 0.5 kg This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
17768949
Encompass_Preferred
View Documents
Minebea Intec™
EF-4PNNS1500-3d
1250 x 1250 x 105 mm Plastic Indicator, Stainless Steel Frame and Load Plate, USB-C Port, 35 mm Weight Display 1500 kg 0.5 kg This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
17778949
Encompass_Preferred
View Documents
Minebea Intec™
EF-4PRRP1500-3d
1500 x 1500 x 105 mm Plastic Indicator, Painted Carbon Steel Frame and Load Plate, USB-C Port, 35 mm Weight Display 1500 kg 0.5 kg This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
17798949
Encompass_Preferred
View Documents
Minebea Intec™
EF-4PRRS1500-3d
1500 x 1500 x 105 mm Plastic Indicator, Painted Carbon Steel Frame, Stainless Steel Load Plate, USB-C Port 1500 kg 0.5 kg This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
17788949
Encompass_Preferred
View Documents
Minebea Intec™
EF-4PRRP3000-3d
1500 x 1500 x 105 mm Plastic Indicator, Painted Carbon Steel Frame and Load Plate, USB-C Port, 35 mm Weight Display 3000 kg 1 kg This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
17708959
Encompass_Preferred
View Documents
Minebea Intec™
EF-4PRRS3000-3d
1500 x 1500 x 105 mm Plastic Indicator, Stainless Steel Frame and Load Plate, USB-C Port, 35 mm Weight Display 3000 kg 1 kg This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
Description
Description
Puro™ floor scales unite the most needed applications in one multi-purpose indicator with large display for a comfortable user operating experience.
  • Accurate and reliable results in a second
  • Combined with the robustness and durability of platforms, floor scales fit comfortably in any environment while offering simple charging and connection technology
  • Up to 500 hours continuous use between recharges with 12 hour recharge time
  • LCD display with white backlight, 35 mm (1.4 in.) height, 6 digits, 14-segment
  • For weighing, counting, check weighing, check counting, and totalization
Specifications
Specifications
Floor Scale
Backlit LCD
IP43 (Indicator), IP67 (Platform)