Learn More
RETSCH Pure 70mm Diameter Stainless Steel Ring Sieve For Ultra Centrifugal Mill ZM 200
Brand:  RETSCH 03.647.0293
 View more versions of this product
Product Code. 11944272
  • 202.00€ / Each
Estimated Shipment: 03-02-2025
to see stock.
Explore more special offers
Specifications
1Each
Ring Sieve
Ultra Centrifugal Mill ZM 200
1.5 mm
Stainless Steel
70 mm